C-4 ≥95% (durch HPLC), Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem

Synonyme: (E)-4-Chloro-N-(3-(3-(4-hydroxy-3-methoxyphenyl)acryloyl)phenyl)benzamide,, P-Glycoprotein Inhibitor

M-Clarity™ Qualitätsstufe = MQ100

517301-10
517301-10EA 199 EUR
517301-10
C-4 ≥95% (durch HPLC), Calbiochem®, Sigma-Aldrich®
C-4

The P-Glycoprotein Inhibitor, C-4 controls the biological activity of P-Glycoprotein. This small molecule/inhibitor is primarily used for Cancer applications.


General description
A cell-permeable cinnamoyl compound that reversibly inhibits P-gp (P-glycoprotein) efflux function and reverses the MDR (multidrug resistance) phenotype with no noticeable effect on P-gp expression. Shown to significantly enhance Paclitaxel- (Cat. No. 580555) and Vincristine- (Cat. No. 677181) induced cytotoxicity as well as apoptosis in P-gp expressing KBV20C cells.


Verpackung: Packaged under inert gas

Formel: C₂₃H₁₈ClNO₄
Molecular Weight: 407,85 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 864426-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR