Dimethylsulfoxid ≥99,9% (durch GC), CHROMASOLV™ für die Headspace-Gaschromatographie, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Achtung

51779-2.5L
51779-2.5LEA 390.74 EUR
51779-2.5L
Dimethylsulfoxid ≥99,9% (durch GC), CHROMASOLV™ für die Headspace-Gaschromatographie, Riedel-de Haën™
Dimethylsulfoxid

Honeywell Chromasolv™ GC-headspace solvents:


  • Developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • Very pure solvents with extremely low concentrations of the defined residual solvents

Formel: (CH₃)₂SO
Siedepunkt: 189 °C
Dichte: 1,104 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002089
CAS-Nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (GC) Min. 99.9 %
Refractive index (n 20/D) 1.477 - 1.480
Water (Karl Fischer) Max. 0.04 %
UV-transmittance at 268 nm Min. 30 %
UV-transmittance at 275 nm Min. 60 %
UV-transmittance at 300 nm Min. 85 %
UV-transmittance at 350 nm Min. 95 %
UV-transmittance from 400 nm Min. 98 %
Suitability for the GC Headspace complying
Vapor Pressure 0.42 mmHg ( 20 °C)
Explosion Limit 42 %, 63 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR