N,N-Dimethylformamid ≥99,9%, CHROMASOLV™ für die Headspace-Gaschromatographie, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: DMF, Ameisensäuredimethylamid, N,N-Dimethylmethansäureamid

51781-1L 51781-6X1L
51781-1LEA 169.17 EUR
51781-1L HONC51781-6X1L
N,N-Dimethylformamid ≥99,9%, CHROMASOLV™ für die Headspace-Gaschromatographie, Riedel-de Haën™
N,N-Dimethylformamid

Honeywell Chromasolv™ GC-headspace solvents:


  • Developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • Very pure solvents with extremely low concentrations of the defined residual solvents

Formel: HCON(CH₃)₂
Molecular Weight: 73.09 g/mol
Siedepunkt: 152…153 °C (1.013 hPa)
Schmelzpunkt: –61 °C
Dichte: 0.94 g/cm³ (20 °C)
Flammpunkt: 57.5 °C (1.013.25 hPa)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003284
CAS-Nummer: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Suitability for the GC Headspace complying
Assay (GC) Min. 99.9 %
Refractive index (n 20/D) 1.429 - 1.431
Water (Karl Fischer) Max. 0.03 %
Absorbance at 270 nm Max. 0.700
Absorbance at 275 nm Max. 0.250
Absorbance at 300 nm Max. 0.070
Absorbance at 320 nm Max. 0.022


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR