Zentrifugalfiltrationseinheiten, Vivaspin® Turbo 4
Lieferant: Sartorius
VS04T01 VS04T02 VS04T11 VS04T12 VS04T21 VS04T22 VS04T31 VS04T32 VS04T41 VS04T42 VS04T91 VS04T92
518-0092EA 266 EUR
518-0092 518-0093 518-0094 518-0095 518-0096 518-0097 518-0098 518-0099 518-0100 518-0101 518-0102 518-0103
Zentrifugalfiltrationseinheiten, Vivaspin® Turbo 4
Filter Zentrifugaleinheiten
Vivaspin® Turbo 4 can handle 0,04 to 4 ml sample volume in swing-out and fixed angle rotors accepting 15 ml centrifuge tubes.
  • High retentate recovery >95%
  • pH 1 to 14 suited PS housing with high chemical compatibility
  • Safe sample concentration and easy recovery due to unique, angular and pipette friendly dead-stop pocket
  • Volume graduation for easy detection/monitoring of the concentrate volume
The Vivaspin® Turbo 4 optimised design, the sleek internal profile, ensures maximum process speeds right the way down to the last few micro litres after >100 fold concentration.