Multi-element standard solution 4 for ICP, TraceCERT®, Supelco®
Lieferant: Merck
51844-100ML
51844-100MLEA
149
EUR
51844-100ML
Multi-element standard solution 4 for ICP, TraceCERT®, Supelco®
Standards
Kationischer Standards
Mehrelement-Standards
17 components
Al, 40 mg/l; As, 40 mg/l; B, 100 mg/l; Ba, 40 mg/l; Be, 10 mg/l
Cd, 10 mg/l; Co, 10 mg/l; Cr, 20 mg/l; Cu, 20 mg/l; Fe, 100 mg/l
Mn, 10 mg/l; Ni, 20 mg/l; Pb, 40 mg/l; Se, 100 mg/l; Tl, 100 mg/l
V, 40 mg/l; Zn, 100 mg/l
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034