Woulff bottles
Lieferant: Sartorius
16610
519-4500EA 167 EUR
519-4500
Woulff bottles
Flaschen Woulffsche Flaschen
Made of borosilicate glass.
  • Side necks with screw caps and inlet/outlet glass tubes
  • Centre neck with screw cap and stopcock
  • Used between suction flask and vacuum source
  • Very good chemical and temperature resistance
With three necks. For protection of vacuum pump.