Supelco® TraceCERT® Organic Reference Standard, Cypermethrin
Lieferant: Merck
- GHS signal: GHS06, GHS08, GHS09
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P273 - P280 - P301 + P312 - P302 + P352 - P304 + P340 + P312 - P314