6-Methoxy-N-(3-sulphopropyl)quinolinium (SPQ) ≥95% (durch HPLC) fluoreszierender Indikator, zur Bestimmung von Chlor
Lieferant: Biotium
|
Achtung
|
Synonyme:
SPQ, 3-(6-Methoxyquinolin-1-ium-1-yl)propane-1-sulphonate
52010
52010.EA
95.3
EUR
52010.
6-Methoxy-N-(3-sulphopropyl)quinolinium (SPQ) ≥95% (durch HPLC) fluoreszierender Indikator, zur Bestimmung von Chlor
6-Methoxy-N-(3-sulphopropyl)quinolinium (SPQ)
SPQ is a blue fluorescent dye (Ex/Em 344/443 nm) that is specifically quenched by chloride via collision (KSV=118 M-1). Therefore, chloride concentrations can be measured by moniotiring the degree of fluorescence decrease. The dye can be loaded into cells by hypotonic shock.
- White solid soluble in H2O and DMSO
- Store at 4°C. Protect from light, especially in solution.
- λEx/λEm= 344nm / 443nm
- ε (344 nm, H2O) = 3,700
Formel:
C₁₅H₁₅NO₄S Molecular Weight: 281 g/mol |
MDL:
MFCD00037592 CAS-Nummer: 83907-40-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...