MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥95% (durch HPLC) fluoreszierender Indikator, zur Bestimmung von Chlor

Lieferant: Biotium
Achtung

Synonyme: (6-Methoxyquinolinio)acetic acid ethyl ester bromide, 1-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide, 1-(2-Ethoxy-2-oxoethyl)-6-methoxyquinolin-1-ium bromide, MQAE

52011
52011.EA 185 EUR
52011.
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥95% (durch HPLC) fluoreszierender Indikator, zur Bestimmung von Chlor
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide)

MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) is an improved chloride indicator that has greater sensitivity to chloride (Ksv= 200M-1) than SPQ (Ksv=118 M-1) and higher fluorescence quantum yield. The ester group of MQAE may slowly hydrolyze inside cells, resulting in a change in its fluorescence response.


  • White solid soluble in water and DMSO
  • Store at 4°C and protect from light, especially in solution
  • λEx/λEm= 350/460nm
  • ε (344 nm, H2O) = 2,800

Formel: C₁₄H₁₆NO₃Br
Molecular Weight: 326,19 g/mol
MDL: MFCD00467854
CAS-Nummer: 162558-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88ca380ec368e65119c76ad9bb3adc7588873cd0&vid=88ca380ec368e65119c76ad9bb3adc7588873cd0&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR