L-α-Phenylalanin, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: H-Phe-OH, H-L-Phe-OH, L-α-Phenylalanin, 3-Phenyl-L(-)-alanin, 3-Phenyl-L-alanin, (S)-(-)-2-Amino-3-phenylpropansäure

5202-100
5202-100EA 130 EUR
5202-100
L-α-Phenylalanin, Millipore®
L-α-Phenylalanin
Formel: C₉H₁₁NO₂
Molecular Weight: 165.19 g/mol
Siedepunkt: 313
Schmelzpunkt: 173
Dichte: 0.754 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064227
CAS-Nummer: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR