Zinquin ethyl ester ≥95% (durch HPLC) fluoreszierender Indikator, für die Bestimmung von Zink
Lieferant: Biotium
Synonyme:
2-[[2-Methyl-8-[[(4-methylphenyl)sulphonyl]amino]-6-quinolinyl]oxy]-acetic acid ethyl ester
52020
52020.EA
352
EUR
52020.
Zinquin ethyl ester ≥95% (durch HPLC) fluoreszierender Indikator, für die Bestimmung von Zink
Zinquin ethyl ester
Zinquin is an UV-excitable, blue fluorescent zinc indicator. Zinquin ethyl ester is membrane-permeable and is hydrolyzed into Zinquin free acid once entering cells. Zinc is believed to be involved in the suppression of apoptosis and thought to play important roles in many neural activities.
- λEx/λEm (with Zn2 )= 364/385 nm
- Cell permeability: Membrane permeant
- Cellular localization: Biotinylated targets
- Colors: Blue
- Off-white solid soluble in DMSO
- Store at 4°C and protect from light
Formel:
C₂₁H₂₂N₂O₅S Molecular Weight: 414,47 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
181530-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...