Zinquin freie Säure ≥98% (durch HPLC) fluoreszierender Indikator, für die Bestimmung von Zink

Lieferant: Biotium

Synonyme: Zinquin A

52022
52022.EA 352 EUR
52022.
Zinquin freie Säure ≥98% (durch HPLC) fluoreszierender Indikator, für die Bestimmung von Zink
Zinquin

Zinquin is an UV-excitable, blue fluorescent zinc indicator. Zinc is believed to be involved in the suppression of apoptosis and thought to play important roles in many neural activities.


  • λEx/λEm (with Zn2 )= 364/385 nm
  • Cell permeability: Membrane impermeant
  • Colors: Blue
  • Off-white solid soluble in DMSO
  • Store at 4°C and protect from light

Formel: C₁₉H₁₈N₂O₅S
Molecular Weight: 386,42 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03452711
CAS-Nummer: 151606-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a056badb1959327c631e49af2fd7d2aa2b2c3d9c&vid=a056badb1959327c631e49af2fd7d2aa2b2c3d9c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR