Xylenolorange Ditriumsalz Spectrophotometry, Supelco®

Lieferant: Merck

Synonyme: 3,3-Bis(N,N-bis(carboxymethyl)aminomethyl)kresolsulfonphthalein

MQ100

52097-1G 52097-5G
52097-1GEA 52.6 EUR
52097-1G 52097-5G
Xylenolorange Ditriumsalz Spectrophotometry, Supelco®
Xylenolorange Ditriumsalz

Reagent for spectrophotometric determination of metal ions

Formel: C₃₁H₃₀N₂Na₂O₁₃S
Molecular Weight: 716.62 g/mol
MDL: MFCD00014580
CAS-Nummer: 1202864-39-8
EINECS: 216-553-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61dbf026e9146752269a2458fb3542cca240e4f7&vid=d8bdd019f78b35e8a24e0c6a1454dcd5e93e0672&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR