Zentrifugen, Tischmodell, ungekühlt/gekühlt, 5702 / 5702 R / 5702 RH (IVD-Version)
Lieferant: EPPENDORF
5702000.019 5703000.012 5704000.016 5702000.060 5703000.063 5704000.067 5702000.329 5703000.322 5702000.361 5703000.365 5703000.420 5703000.462 5703000.489 5703000.381 5702000.510
521-0020EA 2178 EUR
521-0020 521-0042 521-0593 521-0125 521-0134 521-0594 521-0992 521-0993 521-1141 521-1148 521-1718 521-1942 521-2135 521-2134 EPPE5702000.510
Zentrifugen, Tischmodell, ungekühlt/gekühlt, 5702 / 5702 R / 5702 RH (IVD-Version)
Zentrifugen Tischzentrifugen
Die Zentrifugen-Serie 5702 ist ideal für medizinische Routineanwendungen sowie klinische und biotechnologische Forschung.
  • User friendly control panel with a small number of clearly labelled function keys and easy to read LCD display
  • Speed can be set from 100 min⁻¹ to a maximum of 4400 min⁻¹
  • Automatic, motorised lid locking
  • Slow startup and braking through the SOFT function
  • Stainless steel rotor chamber (rust-proof)
  • FastTemp function for fast precooling (5702 R/RH)
  • Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)
The heated and refrigerated centrifuge 5702 RH allows centrifugation under biological conditions to improve cell viability for more accurate results in downstream applications (e.g. cell cultivation). All centrifuges are designed for separating liquid substance mixtures with different densities, in particular, for processing and analysing samples from the human body in in-vitro diagnostic applications to ensure that the in-vitro diagnostic device can be used according to its intended purpose.
There are different rotor options available to accommodate any type of tube. Rotors, buckets and lids are autoclavable. The centrifuges have induction motors and a timer which can be set for up to 90 minutes or for continuous operation.