Rotors for Centrifuge 5430/5430 R
Lieferant: EPPENDORF
5427705.007 5427710.000 5427712.003 5427714.006 5427700.005 5427716.009 5427754.008 5427750.002 5427753.001 5427755.004 5427757.007 5427752.005
521-2013EA 549 EUR
521-2013 521-2039 521-2008 521-2016 521-2010 521-2017 521-1940 521-1941 521-1741 521-2067 521-2068 521-1740
Rotors for Centrifuge 5430/5430 R
Zentrifugen Rotoren
Eppendorf rotors for centrifuge 5430/5430 R, precision at its core. Maximise your centrifuge's potential with rotors crafted for efficiency. Tailor your experiments with various capacities and speeds, ensuring optimal sample processing.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
  • Aerosol-tight and PTFE coated options available
Explore enhanced centrifugation possibilities with Eppendorf rotors designed for centrifuge 5430/5430 R. Choose from a range of capacities and speeds, providing versatility for microcentrifugation or larger sample volumes. The precision-engineered rotors ensure optimal sample separation. Accessories such as adaptors and buckets further customise your centrifuge setup. Maximise efficiency with the ability to handle diverse sample types. Elevate your laboratory's capabilities, achieve reproducible results, and enjoy the flexibility to meet specific application needs.