Rotor assembly, Avanti JS-4.2
Lieferant: Beckman Coulter
339080
521-3705EA 13780 EUR
521-3705
Rotor assembly, Avanti JS-4.2
Zentrifugen Rotoren
Windshielded, six-place rotor with aluminium rotor yoke and removable aluminium swinging buckets. Buckets are interchangeable with microplate carriers (358682) for spinning microtitre plates and minitube racks. Holds up to six litres, six blood bags, or six microtitre plate carriers. Accepts all multi-disc adapters. A kit is available to convert this rotor to a JS-4.2SM (356501)