Rotor assembly, Avanti JS-4.0
Lieferant: Beckman Coulter
339086
521-3706EA 10230 EUR
521-3706
Rotor assembly, Avanti JS-4.0
Zentrifugen Rotoren
Unshielded, four-place rotor with aluminium rotor yoke and removable aluminium swinging buckets. Buckets are interchangeable with microplate carriers for spinning microtitre plates and minitube racks. Holds up to four litres, four blood bags or four microtitre plate carriers. Accepts all multi-disc adapters except 339119. Note: requires tie-down kit (367045) for use with Avanti J-26 XP Series centrifuges.
Bitte beachten NOTE: Requires tie-down kit (367045) for use with Avanti J-26 XP Series centrifuges.