Rotor package, Avanti JLA 9.1000
Lieferant: Beckman Coulter
366754
521-4037EA 21840 EUR
521-4037
Rotor package, Avanti JLA 9.1000
Zentrifugen Rotoren
Empty rotor and windshield weight is 10,8 kg. Package includes: JLA-9.1000 rotor, lid with removable canisters, labware kit (bottle rack, spatula, bottle grip and wrench). Upgrade required for existing instruments prior to 1998.