Fixed-angle titanium rotor package (26° angle), MLA-80
Lieferant: Beckman Coulter
367096
521-4041EA 17160 EUR
521-4041
Fixed-angle titanium rotor package (26° angle), MLA-80
Zentrifugen Rotoren
Major applications: Large volume differential sedimentation (pelleting) of subcellular organelles and viruses.