Floor-standing ultracentrifuge, Optima™ XE-90
Lieferant: Beckman Coulter
A94471
521-4179EA 0 EUR
521-4179
Floor-standing ultracentrifuge, Optima™ XE-90
Zentrifugen Standzentrifugen
The Optima XE has features and beneficial enhancements you will not find anywhere else. With total-system design in mind from the start, the Optima XE has been created to simplify use, increase productivity, and reduce error. Units have an intelligent user interface, multi-layered biosafety features, and energy efficiency. The system design is backed by rotors, labware, and service.
  • Large touchscreen display is easy to read and navigate
  • Remote monitoring and control (XPN only) save time. Start, stop, and monitor from computer or smart phone. Receive diagnostic alerts via e-mail. iPhone or Android compatible
  • Energy efficient, maintains runs during power fluctuations. Regenerative braking returns energy to local circuit. Highly efficient thermoelectric cooling results in lower power consumption. Idle state energy usage equivalent to a 60-Watt light bulb
  • Powerful on-board simulation and calculation tools (XPN only) saves time and samples. Proprietary software optimises protocols before running to ensure maximum efficiency
  • Quickly perform common calculations and conversions
  • Quiet-drive technology, designed for low drive noise. Operates at less than 51 dBA
The visibility and control of the intelligent user interface makes the Optima X Series a breeze to operate. The entry-level model, the XE delivers all the features required to quickly setup and complete a run. The premium model, the XPN, boasts networking capability and a variety of customisable security and tracking features, ensuring chain of custody and compliance. Both models feature multi-layered biosafety features in order to maintain a safe working environment. All instruments are designed as a total system with innovative rotors and labware.
Please see Ultracentrifuge and AUC Product Catalogue section in the PDF catalogue under Documentation for full technical specification.