Sorvall™ MTX and MX+ Micro Ultracentrifuges
Lieferant: Thermo Fisher Scientific
46963 50135643 50135648
521-1956EA 77100 EUR
521-1956 521-4296 521-4295
Sorvall™ MTX and MX+ Micro Ultracentrifuges
Zentrifugen Mikrozentrifugen
Sorvall™ Micro ultracentrifuges, three models offering a maximum RCF of up to 1048000×g in a compact footprint with either bench or floor options.
  • User-friendly LCD touch screen
  • Self locking rotor system
  • Visual tube balancing for enhanced ease of use
  • USB port for connectivity
  • Extensive rotor selection including lightweight, fatigue-resistant Fiberlite™ carbon fibre rotor
Accommodating tubes from 0,2 to 30 ml with a choice of fixed angle, swinging bucket or vertical rotors, theSorvall micro-ultracentrifuges address the full range of small volume processing applications (e.g. virus. Cellular organelles, lipoproteins, nanoparticles and nucleic acids).
Bestellinformation: Supplied without rotors which can be ordered separately.