Pterostilben, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: (E)-4-(3,5-Dimethoxystyryl)phenol, 4-[(E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol

523310-10
523310-10EA 294 EUR
523310-10
Pterostilben, Millipore®
Pterostilben
Formel: C₁₆H₁₆O₃
Molecular Weight: 256.2964 g/mol
MDL: MFCD00238710
CAS-Nummer: 537-42-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR