Wasserstoffperoxid-Lösung 30%, AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Hydrogenperoxid

5240-20EA 0 EUR
5240-20
Wasserstoffperoxid-Lösung 30%, AR® ACS, Macron Fine Chemicals™
Wasserstoffperoxid
Formel: H₂O₂
Molecular Weight: 34.01 g/mol
Siedepunkt: ∼107 °C (1013 hPa)
Schmelzpunkt: <0 °C
Dichte: 1.11 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011333
CAS-Nummer: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 II
Merck Index: 14,04798

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 29.0 - 32.0%
For Laboratory,Research,or Manufacturing Use ≤0.0005%
ACS - Assay (H₂O₂) 29.0 - 32.0 %
ACS - Ammonium (NH₄) ≤0.0005 %
Chloride (Cl) ≤2 ppm
ACS - Color (APHA) ≤10
ACS - Heavy Metals (as Pb) ≤0.0001 %
ACS - Iron (Fe) ≤0.00005 %
ACS - Nitrate (NO₃) ≤0.0002 %
ACS - Phosphate (PO₄) ≤0.0002 %
ACS - Residue after Evaporation ≤0.002 %
ACS - Sulfate (SO₄) ≤0.0005 %
ACS - Titrable Acid (meq/g) ≤0.0006
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR