Phloretin, Millipore®

Lieferant: Merck Millipore Calbiochem

524488-200
524488-200EA 190 EUR
524488-200
Phloretin, Millipore®
Phloretin
Formel: C₁₅H₁₄O₅
Molecular Weight: 274,28 g/mol
MDL: MFCD00002288
CAS-Nummer: 60-82-2
EINECS: 200-488-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR