Supelco® TraceCERT® Organic Reference Standard, Sodium benzoate
Lieferant: Merck
52451-100MG
52451-100MGEA 86.1 EUR
52451-100MG
Supelco® TraceCERT® Organic Reference Standard, Sodium benzoate
Standards Organische Standards
  • GHS signal: GHS07
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P264 - P280 - P305 + P351 + P338 - P337 + P313
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1d1acba51d5ab3e934e23d66e526084945ddfb&vid=8a1d1acba51d5ab3e934e23d66e526084945ddfb&prid=web&clid=SAPDAM