Ammoniumfluorid ≥98,0% Eluentzusatz, für die LC-MS, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Fluoroammonium

52481-50G
52481-50GEA 57.01 EUR
52481-50G
Ammoniumfluorid ≥98,0% Eluentzusatz, für die LC-MS, Fluka™
Ammonium fluoride
Formel: NH₄F
Schmelzpunkt: 100 °C
Dichte: 1,01 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011423
CAS-Nummer: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Assay 98.0 - 102.0 %
Ammonium hydrogen difluoride (NH₄HF₂) Max. 1 %
Residue on ignition (SO₄) Max. 0.005 %
Silver (Ag) Max. 5 ppm
Aluminium (Al) Max. 5 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Molybdenum (Mo) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 5 ppm
Thallium (Tl) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 50 ppm
Suitability for the LC-MS complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR