Centrifuge tube, round bottom, with screw cap, KIMAX®, KIMBLE®
Lieferant: DWK Life Sciences
45212-50 SP45212-50 SP45212-35
KIML45212-50EA 0 EUR
KIML45212-50 525-0788 525-0787
Centrifuge tube, round bottom, with screw cap, KIMAX®, KIMBLE®
Röhrchen Zentrifugenröhrchen
Tube has a heavy wall to withstand higher speeds in centrifuging.
It is especially useful in separating neutralised sediment for culturing from sputum specimens, as well as many other clinical and microbiological applications.
Lieferumfang: Supplied with unattached autoclavable cap with cemented-in rubber liner.