VWR® Ultra-High Performance Centrifuge Tubes
Lieferant: VWR Collection
525-1107EA 263 EUR
525-1107 525-1109 525-1111 525-1113 525-1103 525-1105 525-1097 525-1098 525-1099 525-1100 525-1101 525-1081 525-1082 525-1083 525-1084 525-1085 525-1086 525-1087 525-1088 525-1089 525-1090 525-1091 525-1092 525-1093 525-1094 525-1095 525-1096
VWR® Ultra-High Performance Centrifuge Tubes
Röhrchen Zentrifugenröhrchen
These leakproof, conical bottom centrifuge tubes prevent breakage and leakage during high speed centrifugation, and are made of non cytotoxic, medical grade resin that allows users to easily see sample volume and colour.
  • Easy-to-read black graduations in ±2% increments
  • Contains a large, white frosted writing area
  • RNase-, DNase-, and non-pyrogenic
  • Autoclavable at 121 °C and freezable to –80 °C
  • Sterile tubes are sterilized by irradiation
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use
More Product Information