Centrifuge tubes conical bottom, Corning®
Lieferant: Corning
11750
525-1749EA 276 EUR
525-1749
Centrifuge tubes conical bottom, Corning®
Röhrchen Zentrifugenröhrchen
PP sterile closed system centrifuge tubes arrive individually double bagged and pre-assembled. The caps have two ports: One port ends in a 0,2 µm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in a male Luer. Dip tubes, where available, reach all the way to the bottom of the flask for easy aseptic transfer during your liquid handling process.
These centrifuge tubes are ideal for applications requiring large volume centrifugation.
Each case of tubes contains a recyclable foam rack to facilitate handling. Support cushions must be used with this product unless the centrifuge rotor has appropriate V-bottom cushions. HDPE plug seal caps. Maximum RCF of 6000×g. Tubes are sterile and certified non pyrogenic. Extra cap is included for centrifugation. Conical bottom style with graduations and dip tube. Plug seal cap style.