BROXO Salz, Granulat 6 - 15 mm

Lieferant: VWR Chemicals

Synonyme: Saline, NaCl, Sea salt, Halite

5254.9025EA 62.4 EUR
5254.9025
BROXO Salz, Granulat 6 - 15 mm
Natriumchlorid

Broxo water softener salt allows for problem free water softening that gives a great performance and is efficient in use. It is a very pure high-pressure compacted vacuum salt that dissolves evenly and gradually and is applicable to domestic and industrial installations. Broxo water softener can be used for both domestic and industrial use. The granules dissolves completely and evenly to leave no residual substances. The granules measure from 6mm up to 15mm and provide excellent performance. Also known as rock salt.


  • Very pure high-pressure compacted vacuum salt (99,8% NaCl)
  • For use in domestic and industrial installations
  • Granules from 6 mm up to 15 mm

Formel: NaCl
Molecular Weight: 58.44 g/mol
Siedepunkt: 1413
Schmelzpunkt: 801
Dichte: 2.16 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003477
CAS-Nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR