Melamin, Supelco®
Lieferant: Merck
Synonyme:
2,4,6-Triamino-1,3,5-triazin, 2,4,6-Triamino-s-triazin
M-Clarity™ Qualitätsstufe = MQ100
52549-250MG
52549-250MGEA
33.9
EUR
52549-250MG
Melamin, Supelco®
Melamin
Formel:
C₃H₆N₆ Molecular Weight: 126,12 g/mol Dichte: 1,57 g/cm³ (20 °C) Flammpunkt: >280 °C |
MDL:
MFCD00006055 CAS-Nummer: 108-78-1 EINECS: 203-615-4 Merck Index: 13,05834 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...