Piceatannol, Calbiochem®

Lieferant: Merck Millipore Calbiochem

Synonyme: (E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol

527948-1
527948-1EA 58.8 EUR
527948-1
Piceatannol, Calbiochem®
Piceatannol
Formel: C₁₄H₁₂O₄
Molecular Weight: 244,2427 g/mol
MDL: MFCD00221715
CAS-Nummer: 10083-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR