PIPPS (1,4-Piperazinedipropanesulphonic acid) 97 T-dried, Calbiochem®

Lieferant: Merck Millipore Calbiochem

Synonyme: Piperazine-N,N'-bis(3-propanesulphonic acid), Piperazine-1,4-bis(propanesulphonic acid)

528315-100
528315-100EA 468 EUR
528315-100
PIPPS (1,4-Piperazinedipropanesulphonic acid) 97 T-dried, Calbiochem®
PIPPS (1,4-Piperazinedipropanesulphonic acid)

A non-metal complexing, zwitterionic buffer with mixed-mode dissociation constants of pKa1 = 3.73 and pKa2 = 7.96 (100 mM, 25°C).
PIPPS has no interference at wavelengths longer than 240 nm.


Storage and Shipping Instructions: Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Formel: C₁₀H₂₂N₂O₆S₂
Molecular Weight: 130.093 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00274122
CAS-Nummer: 5625-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR