n-Hexatetracontan, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

52913-250MG
52913-250MGEA 135 EUR
52913-250MG
n-Hexatetracontan, Supelco®
n-Hexatetracontane
Formel: H₃C(CH₂)₄₄CH₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 7098-24-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24e069095ec307d309d561475714013b62fca659&vid=3c1ce0cc8258300600af79a7ad33daba7af0186e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR