Flasks, round bottom, glass, clear, with spherical joints
Lieferant: witeg Labortechnik
0.645.250 0.645.500 0.645.001 0.645.002
531-9133EA 66.8 EUR
531-9133 531-9134 531-9135 531-9136
Flasks, round bottom, glass, clear, with spherical joints
Kolben Rundkolben
Borosilicate glass 3.3, plastic coated.
  • Very good chemical resistance
  • Good temperature resistance