Vacuum seal PTFE / FKM
Lieferant: Heidolph Instruments GmbH & Co.KG
23-30-01-01-30
531-9423EA 59 EUR
531-9423
Vacuum seal PTFE / FKM
Versiegeln Dichtringe
The highly durable and long lasting PTFE vacuum seals with FKM sealing ring achieve maximum tightness and save on long-term spare parts costs. The combination of both materials allows for very good chemical resistance, while at the same time offering the best sealing properties.
  • Standard PTFE spare seal (with FKM sealing ring)
  • Particularly high chemical resistance
  • Long running time without abrasion
  • Highest density