Scheidetrichter, birnenförmig, Quickfit®
Lieferant: SciLabware
D2/01 D2/11 D2/21 D2/22 D2/32 D2/42 D2/43
532-0079EA 102 EUR
532-0079 532-0080 532-0082 532-0083 532-0085 532-0086 532-0087
Scheidetrichter, birnenförmig, Quickfit®
Trichter Scheidetrichter
PYREX®, borosilicate glass.
  • Without graduation
  • Excellent chemical resistance
  • Excellent temperature resistance
Funnel stem with cone to allow easy connection to Quickfit® flask with interchangeable PTFE key stopcock.
Standards: Complies with ISO 4800 type 2.