Automated Kjeldahl steam distillation units
Lieferant: SELECTA
4002851
533-0047EA 10700 EUR
533-0047
Automated Kjeldahl steam distillation units
Destillationssysteme Kjeldahl-Destillationssysteme
A range of compact distillation units for total nitrogen and protein analysis according to Kjeldahl, with the reservoirs for the H₂O and reagents located within the units. Three models are available depending on the level of sophistication and automation required. All the units have the same basic features including.
  • Compact steam generator with safety over temperature and over pressure device
  • Safety door - the system will not operate if the door is open
  • Tube in place sensor – if tube not located then dosing process for NaOH is prevented
  • Universal adapter available, for macro and micro distillation tubes
  • Stainless steel case with reinforced ABS plastic front
Pro–Nitro M: Basic distillation unit for small to medium throughput of samples. Mainly manual features with limited automation.
Pro–Nitro S: Semi automatic distillation unit for medium to large throughput of samples. Choice of manual or automatic functionality.
Pro–Nitro A: Automated ‘on-line’ analysis of the distillate allows for systematic, safe, precise analysis with minimal personnel intervention. The automatic detection of the end of the process by titration minimises the time cycle and improves accuracy and throughput efficiency. Results can be downloaded to an external printer (optional) for GLP.
Bestellinformation: For complete Kjeldahl digestion and analysis a digestion heating block is reqiuired. Please ask for details. For micro distillation tubes (26 mm 100 ml), please ask for details.
Lieferumfang: Instruments supplied with a MACRO (Ø 42 mm, 250 ml) tube, set of reservoirs and connection tubes. The Pro-Nitro-A also is supplied with 5 litres of boric acid solution and a titration indicator.