Aceton VLSI für Mikroelektronik, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
5356-05EA
113
EUR
5356-05
Aceton VLSI für Mikroelektronik, J.T.Baker®
Aceton
Formel:
CH₃COCH₃ Siedepunkt: 56,2 °C (1013 hPa) Schmelzpunkt: –95,4 °C Dichte: 0,792 g/cm³ (20 °C) Flammpunkt: <–20 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008765 CAS-Nummer: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II REACH: 01-2119471330-49 Merck Index: 13,00067 |
|
Spezifikation Testergebnisse
For Microelectronic Use |
|
Assay ((CH₃)₂CO) (by GC, corrected for water) | ≥99.5 % |
Color (APHA) | ≤10 |
Residue after Evaporation | ≤5 ppm |
Acidity (µeq/g) | ≤0.3 |
Alkalinity (µeq/g) | ≤0.5 |
Water (H₂O) | ≤0.5 % |
Solubility in H₂O | Passes Test |
Chloride (Cl) | ≤0.2 ppm |
Phosphate (PO₄) | ≤0.05 ppm |
Trace Impurities - Aluminum (Al) | ≤50.0 ppb |
Arsenic and Antimony (as As) | ≤5.0 ppb |
Trace Impurities - Barium (Ba) | ≤20.0 ppb |
Trace Impurities - Boron (B) | ≤20.0 ppb |
Trace Impurities - Calcium (Ca) | ≤25.0 ppb |
Trace Impurities - Chromium (Cr) | ≤20.0 ppb |
Trace Impurities - Copper (Cu) | ≤10.0 ppb |
Trace Impurities - Gold (Au) | ≤20 ppb |
Trace Impurities - Iron (Fe) | ≤20.0 ppb |
Trace Impurities - Lead (Pb) | ≤20.0 ppb |
Trace Impurities - Lithium (Li) | ≤50.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤20 ppb |
Trace Impurities - Manganese (Mn) | ≤10.0 ppb |
Trace Impurities - Nickel (Ni) | ≤10.0 ppb |
Trace Impurities - Potassium (K) | ≤50 ppb |
Trace Impurities - Sodium (Na) | ≤50.0 ppb |
Trace Impurities - Tin (Sn) | ≤50 ppb |
Trace Impurities - Titanium (Ti) | ≤20 ppb |
Trace Impurities - Zinc (Zn) | ≤50 ppb |
Particle Count - 0.5 µm and greater | ≤60 par/ml |
Particle Count - 1.0 µm and greater | ≤8 par/ml |
Reported value is the average of all samples counted for this lot number,with no individual sample value exceeding the specification. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...