L(-)-Prolin, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: H-Pro-OH, (S)-(-)-Azolidin-2-carbonsäure, (S)-(-)-1,3,4,5-Tetrahydro-2H-pyrrol- 2-carbonsäure, (S)-(-)-Pyrrolidin-2-carbonsäure, (2S)-(-)-Pyrrolidin-2-carbonsäure, (S)-(-)-Prolin

5370-100
5370-100EA 130 EUR
5370-100
L(-)-Prolin, Millipore®
L(-)-Prolin
Formel: C₅H₉NO₂
Molecular Weight: 115.13 g/mol
Siedepunkt: 245
Schmelzpunkt: 220…222 °C
Dichte: 1.35 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064318
CAS-Nummer: 147-85-3
EINECS: 205-702-2
Merck Index: 13,07871

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR