ortho-Phosphorsäure ≥85% VLSI für die Elektronikindustrie, J.T.Baker®

Lieferant: Avantor
Gefahr

5372-05EA 415 EUR
5372-05
ortho-Phosphorsäure ≥85% VLSI für die Elektronikindustrie, J.T.Baker®
ortho-Phosphorsäure
Formel: H₃PO₄
Siedepunkt: ∼158 °C (1013 hPa)
Schmelzpunkt: ∼21 °C
Dichte: 1,541 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011340
CAS-Nummer: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III
REACH: 01-2119485924-24
Merck Index: 14,07344

Bestellen

Spezifikation Testergebnisse

For Microelectronic Use
Store at temperatures above 22°C (72°F)
Assay (H₃PO₄) (by acidimetry) 85.0 - 87.0 %
Color (APHA) ≤10
Specific Gravity at 60°/60°F 1.691 - 1.710
Reducing Substances Passes Test
Volatile Acids (µeq/g) ≤0.16
Chloride (Cl) ≤1.0 ppm
Nitrate (NO₃) ≤2 ppm
Sulfate (SO₄) ≤12 ppm
Trace Impurities - Aluminum (Al) ≤0.500 ppm
Trace Impurities - Antimony (Sb) ≤10.000 ppm
Trace Impurities - Arsenic (As) ≤0.050 ppm
Trace Impurities - Calcium (Ca) ≤1.500 ppm
Trace Impurities - Chromium (Cr) ≤0.200 ppm
Trace Impurities - Cobalt (Co) ≤0.050 ppm
Trace Impurities - Copper (Cu) ≤0.050 ppm
Trace Impurities - Gold (Au) ≤0.300 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Trace Impurities - Lead (Pb) ≤0.300 ppm
Trace Impurities - Lithium (Li) ≤0.100 ppm
Trace Impurities - Magnesium (Mg) ≤0.20 ppm
Trace Impurities - Manganese (Mn) ≤0.100 ppm
Trace Impurities - Nickel (Ni) ≤0.200 ppm
Trace Impurities - Potassium (K) ≤1.500 ppm
Trace Impurities - Sodium (Na) ≤2.500 ppm
Trace Impurities - Strontium (Sr) ≤0.100 ppm
Trace Impurities - Titanium (Ti) ≤0.300 ppm
Trace Impurities - Zinc (Zn) ≤2.000 ppm
Particle Count - 0.5 µm and greater ≤150 par/ml
Particle Count - 1.0 µm and greater ≤10 par/ml
Reported value is the average of all samples counted for this lot number
with no individual sample value exceeding the specification.
For additional information, go to www.askavantor.com. Search keywords
freezing" and product name."


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR