4-Methyl-2-pentanon VLSI für Mikroelektronik, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK

5384-03EA 233 EUR
5384-03
4-Methyl-2-pentanon VLSI für Mikroelektronik, J.T.Baker®
4-Methyl-2-pentanon
Formel: (CH₃)₂CHCH₂COCH₃
Molecular Weight: 100,16 g/mol
Siedepunkt: 117,4 °C (1013 hPa)
Schmelzpunkt: –84 °C
Dichte: 0,801 g/cm³ (20 °C)
Flammpunkt: 16 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008938
CAS-Nummer: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Microelectronic Use Passes Test
Appearance Passes Test
Assay (CH₃COCH₂CH(CH₃)₂) (by GC, corrected for water) ≥98.5 %
Color (APHA) ≤15
Density (g/mL) at 25°C 0.794 - 0.798
Residue after Evaporation ≤0.0050 %
Acidity (as CH₃COOH) ≤0.01 %
Water (H₂O)(by Karl Fischer titrn) ≤0.1 %
Particle Count - 0.5 µm and greater ≤130 par/ml
Particle Count - 1.0 µm and greater ≤10 par/ml
Boiling Range (Initial) ≥114.0 °C
Boiling Range (Dry Point) ≤117.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR