Organic reference standard, 2,6-Dimethoxyphenol, analytical standard, Supelco®
Lieferant: Merck
53877-100MG
53877-100MGEA
125
EUR
53877-100MG
Organic reference standard, 2,6-Dimethoxyphenol, analytical standard, Supelco®
Standards
Organische Standards
2,6-Dimethoxyphenol can find applications mainly as a substrate of laccases catalysis and also determine its activity in aqueous solution. It can also be used as a potential marker compound for emission of smoke in the atmosphere from wood.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.