Organic reference standard, 2,6-Dimethoxyphenol, analytical standard, Supelco®
Lieferant: Merck
53877-100MG
53877-100MGEA 125 EUR
53877-100MG
Organic reference standard, 2,6-Dimethoxyphenol, analytical standard, Supelco®
Standards Organische Standards
2,6-Dimethoxyphenol can find applications mainly as a substrate of laccases catalysis and also determine its activity in aqueous solution. It can also be used as a potential marker compound for emission of smoke in the atmosphere from wood.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9c1a9c76399b820ff30690d18a504c102fd3e20&vid=d9c1a9c76399b820ff30690d18a504c102fd3e20&prid=web&clid=SAPDAM