Organic reference standard, disperse orange 3, analytical standard, Supelco®
Lieferant: Merck
53882-25MG
53882-25MGEA
165
EUR
53882-25MG
Organic reference standard, disperse orange 3, analytical standard, Supelco®
Standards
Organische Standards
Used to provide experimental verification of a two-level model for the enhancement of the nonresonant, nonlinear refractive index, with a decrease in absorption by reduction of a donor group strength.