Chloroform, AR®, Laborreagens, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

5411-45EA 484.33 EUR
5411-45 5411-25
Chloroform, AR®, Laborreagens, Macron Fine Chemicals™
Chloroform
Formel: CHCl₃
MDL: MFCD00000826
CAS-Nummer: 67-66-3
UN: 1888
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR