Cyclohexan, AR®, Laborreagens, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
5412-25EA
155.75
EUR
5412-25
Cyclohexan, AR®, Laborreagens, Macron Fine Chemicals™
Cyclohexan
Formel:
C₆H₁₂ |
MDL:
MFCD00003814 CAS-Nummer: 110-82-7 UN: 1145 ADR: 3,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...