2-(4-Hydroxyphenylazo)benzoesäure, Supelco®

Lieferant: Merck

Synonyme: 4-Hydroxyazobenzene-2'-carboxylic acid, 2-(4-Hydroxyphenylazo)benzoic acid, 4'-Hydroxyazobenzene-2-carboxylic acid, 2-((4-Hydroxyphenyl)diazenyl)benzoic acid, HABA

M-Clarity™ Qualitätsstufe = MQ100

54793-1G-F
54793-1G-FEA 44.4 EUR
54793-1G-F
2-(4-Hydroxyphenylazo)benzoesäure, Supelco®
2-(4-Hydroxyphenylazo)benzoesäure
Formel: C₁₃H₁₀N₂O₃
Molecular Weight: 242,23 g/mol
MDL: MFCD00002428
CAS-Nummer: 1634-82-8
EINECS: 216-655-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53036d5af6c63d77d5b2894cca09cd4ecf018382&vid=a6890523425e9fb6a6d28d8f0e5688d9f5f853e0&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR