Storage vials, without caps, Chromacol™
Lieferant: CHROMACOL
3.5-HRSV
548-0687EA 1590 EUR
548-0687
Storage vials, without caps, Chromacol™
Vials
These vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass and feature a 13-425 thread finish.