Chromatographieampullen-Verschlüsse, Screw caps with septa, closed top, Härte: 60° shore A, Septum: Red-orange natural rubber/transparent TEF, Stärke: 1,3 mm
Lieferant: VWR Collection
Chromatographieampullen-Verschlüsse, Screw caps with septa, closed top, Härte: 60° shore A, Septum: Red-orange natural rubber/transparent TEF, Stärke: 1,3 mm