Vial racks, Chromacol™
Lieferant: CHROMACOL
T-15/302 T-15/308 T-180 T-28
548-1322EA 0 EUR
548-1322 549-0182 548-1324 548-1327
Vial racks, Chromacol™
Gestelle Gestelle für Chromatographie-Ampullen
These vial racks are chemically resistant and fairly robust for a safe working position on the lab bench and during transport.
  • Ideal for space saving storage in fridges
  • Temperature resistant from −90 to 121 °C (except foam)